missing translation for 'onlineSavingsMsg'
Learn More

Ethacrynic Acid-d5, TRC

Product Code. 17917699
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17917699 10 mg 10mg
17988519 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17917699 Supplier Toronto Research Chemicals Supplier No. TRCE67600210MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330052-59-9
Molecular Formula C13H7D5Cl2O4
Molecular Weight (g/mol) 308.17
Synonym 2-[2,3-Dichloro-4-(2-methylene-1-oxobutyl)phenoxy-d5]acetic Acid, [4-(2-Methylenebutyryl)-2,3-dichlorophenoxy-d5]acetic Acid, Crinuril-d5, Edecril-d5, Edecrin-d5, Edecrina-d5, Endecril-d5, Etacrynic Acid-d5, Etakrinic Acid-d5, Hidromedin-d5, Hydromedin-d5, MK 595-d5, Mingit-d5, NSC 624008-d5, NSC 85791-d5, Otacril-d5, Reomax-d5, Taladren-d5, Uregit-d5;
SMILES ClC1=C(Cl)C(OCC(O)=O)=CC=C1C(C(C([2H])([2H])C([2H])([2H])[2H])=C)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H12Cl2O4/c1-3-7(2)13(18)8-4-5-9(12(15)11(8)14)19-6-10(16)17/h4-5H,2-3,6H2,1H3,(H,16,17)/i1D3,3D2
Chemical Name or Material Ethacrynic Acid-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.