missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic acid, 97%, Thermo Scientific™

Product Code. 11357645
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357645 50 g 50g
11341567 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357645 Supplier Thermo Scientific Alfa Aesar Supplier No. L02270.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethanesulfonic acid is used as pharmaceutical intermediates, catalyst of polymerization and alkylation. It participates in electrolytic reduction of perrhenate solutions.

Solubility
Completely soluble in water.

Notes
Store at room temperature. Incompatible with Strong oxidizing agents, Strong bases. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-45-6
Molecular Formula C2H5O3S
Molecular Weight (g/mol) 109.12
MDL Number MFCD00007529
InChI Key CCIVGXIOQKPBKL-UHFFFAOYSA-M
Synonym ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
PubChem CID 11668
ChEBI CHEBI:42465
IUPAC Name ethanesulfonic acid
SMILES CCS([O-])(=O)=O

Specifications

Melting Point -17°C
Boiling Point 122°C to 123°C (0.01 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
UN Number UN2586
Beilstein 1743071
Formula Weight 110.13
Percent Purity 97%
Chemical Name or Material Ethanesulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.