missing translation for 'onlineSavingsMsg'
Learn More

Ethanethiol, TRC

Product Code. 30369017
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369017 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369017 Supplier Toronto Research Chemicals Supplier No. TRCE676470500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanethiol
CAS 75-08-1
Purity Grade Notes GC
Molecular Formula C2 H6 S
Quantity 500 mL
Synonym Ethyl hydrosulfide, Ethyl mercaptan, Ethyl thioalcohol, Mercaptoethane, NSC 93877, Thioethanol, Thioethyl alcohol
InChI Formula InChI=1S/C2H6S/c1-2-3/h3H,2H2,1H3
SMILES CCS
IUPAC Name ethanethiol
Molecular Weight (g/mol) 62.134
Formula Weight 62.019
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.