missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 200mg/100mL in Water, LoGiCal

Product Code. 30463484
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463484 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463484 Supplier LoGiCal Supplier No. LGCETH200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Ethanol 200mg/100mL in Water
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanol
CAS 64-17-5
Quantity 1 mL
Molecular Formula C2 H6 O
Formulation Single Solution
Synonym Ethanol, Dehydrated Alcohol, Ethanol (9 CI, ACI), Ethyl alcohol (6 CI, 7 CI, 8 CI), 100 C.NPA, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A
InChI Formula InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Formula Weight 46.0419 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.