missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 25mg/100mL in Water, LoGiCal

Product Code. 30461437
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461437 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461437 Supplier LoGiCal Supplier No. LGCETH25

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Ethanol 25mg/100mL in Water
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanol
CAS 64-17-5
Quantity 1 mL
Molecular Formula C2 H6 O
Formulation Single Solution
Synonym Ethanol, Dehydrated Alcohol, Ethanol (9 CI, ACI), Ethyl alcohol (6 CI, 7 CI, 8 CI), 100 C.NPA, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A, SDA 3 A
InChI Formula InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Formula Weight 46.0419 g/mol
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.