missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Denatured with Eurodenaturat, (Disolol™) For Laboratory Use, Chem Lab

Product Code. 16255292
Change view
Click to view available options
Quantity:
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16255292 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16255292 Supplier Chem-Lab Supplier No. CL00.1807.5000
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point -117°C
CAS 64-17-5
Color Colorless
Density 0.79 g/mL
Quantity 5 L
Flash Point 12 °C
Packaging Polyethylene
Physical Form Liquid
Assay Percent Range > 99%
Molecular Formula C2H6O
Water < 0.1%
Boiling Point 78°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Show More Show Less
compliance-icons
Product Identifier
  • Disolol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: disolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.