missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethoxyquin

Product Code. 16660832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16660832 100 g 100g
16650832 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16660832 Supplier Thermo Scientific Chemicals Supplier No. 255631000
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-53-2 | C14H19NO | 217.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-53-2
Molecular Formula C14H19NO
Molecular Weight (g/mol) 217.31
InChI Key DECIPOUIJURFOJ-UHFFFAOYSA-N
IUPAC Name 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline
SMILES CCOC1=CC=C2NC(C)(C)C=C(C)C2=C1

Specifications

Boiling Point 161.0°C (1.6 kPa)
Flash Point 137°C
Quantity 100 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very soluble in methanol, acetone, chloroform,, ethanol, toluene and hexane
Formula Weight 217.31
Chemical Name or Material Ethoxyquin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.