missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyquin, ≥90%, MP Biomedicals™

Product Code. 11445411 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445411 1 kg 1kg
11425411 100 g 100g
11435411 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445411 Supplier MP Biomedicals Supplier No. 0215796391
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 91-53-2
Molecular Formula C14H19NO
Molecular Weight (g/mol) 217.312
MDL Number MFCD00023883
InChI Key DECIPOUIJURFOJ-UHFFFAOYSA-N
Synonym ethoxyquin, 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, ethoxyquine, santoquin, santoquine, niflex, antioxidant ec, santoflex a, santoflex aw, stop-scald
PubChem CID 3293
ChEBI CHEBI:77323
IUPAC Name 6-ethoxy-2,2,4-trimethyl-1H-quinoline
SMILES CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C

Specifications

Color Brown
Density 1.030g/mL
Assay Percent Range ≥90%
Quantity 1 kg
Formula Weight 217.3
Physical Form Viscous Liquid
Chemical Name or Material Ethoxyquin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.