missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyquin, TRC

Product Code. 30137550
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
Unit Size:
100mg
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137550 10 g 10g
30136492 25 g 25g
17928169 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137550 Supplier Toronto Research Chemicals Supplier No. TRCE89280010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-53-2
Molecular Formula C14 H19 N O
Molecular Weight (g/mol) 217.31
Synonym 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 1,2-Dihydro-2,2,4-trimethyl-6-ethoxyquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 2,2,4-Trimethyl-1,2-dihydro-6-ethoxyquinoline, 2,2,4-Trimethyl-6-ethoxy-1,2-dihydroquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, AW, Amea 100, Antage AW, Antioxidant AW, Antioxidant EC, Antox, Aries Antox, BIOX-S, Biox SE, Dawe's nutrigard, EMQ, EQ, Ethoxyquin, Ethoxyquine, NSC 6795, Niflex, Niflex D, Nocrac AW, Nocrac AW-G, Nonflex AW, Nonflex AW-P, Permanax 103, Quinol ED, Raluquin, Santoflex A, Santoflex AW, Santoquin, Santoquine, Stop-Scald, Tenox II
IUPAC Name 6-ethoxy-2,2,4-trimethyl-1H-quinoline
SMILES CCOc1ccc2NC(C)(C)C=C(C)c2c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H19NO/c1-5-16-11-6-7-13-12(8-11)10(2)9-14(3,4)15-13/h6-9,15H,5H2,1-4H3
Formula Weight 217.1467
Percent Purity >95%
Chemical Name or Material Ethoxyquin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.