missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(4-Methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate, TRC

Product Code. 30369019
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369019 100 mg
30370599 1 g
30380820 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369019 Supplier Toronto Research Chemicals Supplier No. TRCE679570100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536759-91-8
Molecular Formula C22 H20 N4 O6
Molecular Weight (g/mol) 436.417
Synonym 4,5,6,7-Tetrahydro-1-(4-methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid Ethyl Ester
IUPAC Name ethyl 1-(4-methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate
SMILES CCOC(=O)c1nn(c2ccc(OC)cc2)c3C(=O)N(CCc13)c4ccc(cc4)[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H20N4O6/c1-3-32-22(28)19-18-12-13-24(14-4-6-16(7-5-14)26(29)30)21(27)20(18)25(23-19)15-8-10-17(31-2)11-9-15/h4-11H,3,12-13H2,1-2H3
Formula Weight 436.138
Chemical Name or Material Ethyl 1-(4-Methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.