missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl(1-hexadecyl)dimethylammonium bromide, 99%

Product Code. 11426886
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
Product Code. Quantity unitSize
11426886 25g 25g
1 options
This item is not returnable. View return policy

Product Code. 11426886

Brand: Alfa Aesar™ B23991.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 124-03-8
Molecular Formula C20H44BrN
MDL Number MFCD00011774
Quantity 25g
Beilstein 3729134
Merck Index 14,2029
Synonym cetyldimethylethylammonium bromide, ethylhexadecyldimethylammonium bromide, cetylcide, bretol, ethyl cetab, ammonyx dme, cetethyldimonium bromide, n-ethyl-n,n-dimethylhexadecan-1-aminium bromide, dimethylethylhexadecylammonium bromide, quaternium 17
InChI Key VUFOSBDICLTFMS-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]
IUPAC Name ethyl-hexadecyl-dimethylazanium;bromide
Molecular Weight (g/mol) 378.483
PubChem CID 31280
Formula Weight 378.49
Percent Purity 99%
Chemical Name or Material Ethyl(1-hexadecyl)dimethylammonium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.