missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-2,2,2-d3-triphenylphosphonium Bromide, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Ethyltriphenylphosphonium-D3 (ethyl-2,2,2-D3) Bromide |
| CAS | 1560-55-0 |
| Molecular Formula | C20 D3 H17 P . Br |
| Quantity | 1 g |
| Synonym | Phosphonium, 1-(ethyl-2,2,2-d3)-1,1,1-triphenyl-, bromide (1:1) (ACI), Ethyl-2,2,2-d3-triphenylphosphonium bromide (7 CI), Phosphonium, ethyl-2,2,2-d3-triphenyl-, bromide (9 CI), (2,2,2-Trideuteroethyl)triphenylphosphonium bromide, ([2-2H3]Ethyl)triphenylphosphonium bromide, Ethyltriphenylphosphonium-D3 (ethyl-2,2,2-D3) bromide |
| InChI Formula | InChI=1 S/C20H20P.BrH/c1-2-21(18-12-6-3-7-13-18,19-14-8-4-9-15-19)20-16-10-5-11-17-20;/h3-17 H,2H2,1H3;1 H/q+1;/p-1/i1D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])C[P+](c1ccccc1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | triphenyl(2,2,2-trideuterioethyl)phosphanium;bromide |
| Molecular Weight (g/mol) | 374.2691 |
| Formula Weight | 373.0674 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction