missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-Dichloro-5-pyrimidinecarboxylate, 97%, BLD Pharm™

Product Code. 31003005
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31003005 25 g 25g
31001895 1 g 1g
30992047 5 g 5g
30992319 500 g 500g
30993311 100 g 100g
31001898 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31003005 Supplier BLD Pharm Supplier No. BD12807225g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 51940-64-8
Molecular Formula C7H6Cl2N2O2
MDL Number MFCD09910281
Quantity 25 g
SMILES C1=C(C(=NC(=N1)Cl)Cl)C(=O)OCC
Molecular Weight (g/mol) 221.04
Percent Purity 97%
Chemical Name or Material Ethyl 2,4-Dichloro-5-pyrimidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.