Learn More
Ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate, 95%, Thermo Scientific™
Product Code. 15438665
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15438665
Brand: Thermo Scientific Alfa Aesar H33837.03
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78185-92-9
Molecular Formula C7H9F6NO2
Molecular Weight (g/mol) 253.14
MDL Number MFCD00040845
InChI Key PGOSQXOLEQZPBM-UHFFFAOYNA-N
Synonym ethyl 4,4,4,4',4',4'-hexafluoro-dl-valinate, ethyl 2-amino-4,4,4-trifluoro-3-trifluoromethyl butanoate, ethyl 2s-2-amino-4,4,4-trifluoro-3-trifluoromethyl butanoate, 4,4,4,4',4',4'-hexafluorovaline, ethyl ester, ethyl 2-amino-4,4,4-trifluoro-3-trifluoromethyl b, ethyl 2-amino-4,4,4-trifluoro-3-trifluoromethyl butyrate, 2-amino-4,4,4-trifluoro-3-trifluoromethyl-butyric acid ethyl ester
PubChem CID 52991940
IUPAC Name ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoate
SMILES CCOC(=O)C(N)C(C(F)(F)F)C(F)(F)F
Specifications
Boiling Point 70°C to 72°C (10 mmHg)
Refractive Index 1.36
Quantity 1 g
Formula Weight 253.14
Percent Purity 95%
Chemical Name or Material Ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate