missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (±)-2-Bromopropionate-3,3,3-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-Bromopropanoate-3,3,3-D3 |
| CAS | 1398066-14-2 |
| Molecular Formula | C5 D3 H6 Br O2 |
| Quantity | 0.25 g |
| Synonym | Ethyl (±)-2-Bromopropionate-3,3,3-d3, Ethyl 2-bromopropanoate-3,3,3-D3, Ethyl 2-bromopropionate-d3, Ethyl α-bromopropionate-β-d3, 2-Bromopropanoic-3,3,3-d3 acid ethyl ester, 2-Bromopropionic-3,3,3-d3 acid ethyl ester |
| InChI Formula | InChI=1 S/C5H9BrO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Br)C(=O)OCC |
| IUPAC Name | ethyl 2-bromo-3,3,3-trideuteriopropanoate |
| Molecular Weight (g/mol) | 184.0463 |
| Formula Weight | 182.9974 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction