missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromovalerate, TRC

Product Code. 30133460
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133460 10 g 10g
30134633 25 g 25g
30005574 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133460 Supplier Toronto Research Chemicals Supplier No. TRCE90055010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-83-8
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.081
Synonym Ethyl α-Bromovalerate, NSC 8865, 2-Bromopentanoic Acid Ethyl Ester;
IUPAC Name ethyl 2-bromopentanoate
SMILES CCCC(Br)C(=O)OCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H13BrO2/c1-3-5-6(8)7(9)10-4-2/h6H,3-5H2,1-2H3
Formula Weight 208.0099
Chemical Name or Material Ethyl 2-Bromovalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.