missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-fluoroacetoacetate, 97%, Thermo Scientific™

Product Code. 10051284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051284 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10051284 Supplier Thermo Scientific Acros Supplier No. 378440250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1522-41-4
Color Colorless to Yellow
Density 1.1810g/mL
Boiling Point 183°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H9FO3
Linear Formula CH3COCH(F)COOCH2CH3
MDL Number MFCD00215830
Quantity 25 mL
Specific Gravity 1.181
Synonym ethyl 2-fluoroacetoacetate, ethyl2-fluoroacetoacetate, 2-fluoro-3-oxo-butanoic acid ethyl ester, 2-fluoroacetoacetic acid ethyl ester, ethyl 2-fluoro-3-oxobutyrate, 2-fluoro-3-oxo-butyric acid ethyl ester, ethyl 2-fluoro-3-oxo-butanoate, ethyl-2-fluoroacetoacetate, ethyl-2-fluoro-3-oxobutanoate, fluoroacetoacetic acid ethyl ester
Solubility Information Solubility in water: soluble
InChI Key SHTFQLHOTAJQRJ-UHFFFAOYSA-N
SMILES CCOC(=O)C(C(=O)C)F
IUPAC Name ethyl 2-fluoro-3-oxobutanoate
Molecular Weight (g/mol) 148.13
PubChem CID 230078
Formula Weight 148.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-fluoroacetoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-fluoroacetoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.