missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (±)-2-Methylbutyrate-d9, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methylbutanoic Acid Ethyl Ester-D9 (2-methylbutanoic-D9) |
| CAS | 2708286-78-4 |
| Molecular Formula | C7 D9 H5 O2 |
| Quantity | 0.25 g |
| Synonym | Ethyl (±)-2-Methylbutyrate-d9, 2-Methylbutanoic-d9 acid ethyl ester, Ethyl 2-methylbutanoate-d9, Ethyl 2-(methyl-d3)butyrate-2,3,3,4,4,4-d6, 2-(Methyl-d3)butanoic-2,3,3,4,4,4-d6 Acid Ethyl Ester, 2-Methylbutanoic Acid Ethyl Ester-D9 (2-methylbutanoic-D9) |
| InChI Formula | InChI=1 S/C7H14O2/c1-4-6(3)7(8)9-5-2/h6H,4-5H2,1-3H3/i1D3,3D3,4D2,6 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)OCC)C([2 H])([2 H])[2 H] |
| IUPAC Name | ethyl 2,3,3,4,4,4-hexadeuterio-2-(trideuteriomethyl)butanoate |
| Molecular Weight (g/mol) | 139.2403 |
| Formula Weight | 139.1559 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction