missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-methylphenoxy)acetate, 99%, Thermo Scientific™

Product Code. 11460477
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460477 100 g 100g
11440477 5 g 5g
11450477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460477 Supplier Thermo Scientific Alfa Aesar Supplier No. B22375.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93917-68-1
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00216559
InChI Key NHFQSFIBSJPLIW-UHFFFAOYSA-N
Synonym ethyl 2-methylphenoxy acetate, ethyl 2-2-methylphenoxy acetate, ethyl 2-o-tolyloxy acetate, ethyl 2-methylphenoxyacetate, ethyl-2-methylphenoxy acetate, o-tolyloxyacetic acid ethyl ester, o-tolyloxy-acetic acid ethyl ester, ethyl 2-2-methylphenoxy ethanoate
PubChem CID 3022879
IUPAC Name ethyl 2-(2-methylphenoxy)acetate
SMILES CCOC(=O)COC1=CC=CC=C1C

Specifications

Density 1.073
Boiling Point 119°C to 120°C
Flash Point >110°C (230°F)
Refractive Index 1.503
Quantity 100 g
Formula Weight 194.23
Percent Purity 99%
Chemical Name or Material Ethyl (2-methylphenoxy)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.