missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxo-4-phenylbutyrate, 90+%

Product Code. 11420691
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420691 50 g 50g
11410691 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420691 Supplier Thermo Scientific Alfa Aesar Supplier No. B20148.18
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64920-29-2 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64920-29-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00037533
InChI Key STPXIOGYOLJXMZ-UHFFFAOYSA-N
Synonym ethyl 2-oxo-4-phenylbutyrate, ethyl benzylpyruvate, ethyl-2-oxo-4-phenylbutyrate, unii-5ynu9lpt51, 5ynu9lpt51, 2-oxo-4-phenylbutyric acid ethyl ester, 2-oxo-4-phenyl butyric acid ethyl ester, ethyl 2-oxo-4-phenyl-butanoate, 2-oxo-4-phenylbutanoic acid ethyl ester, ethyl2-oxo-4-phenylbutyrate
PubChem CID 562087
IUPAC Name ethyl 2-oxo-4-phenylbutanoate
SMILES CCOC(=O)C(=O)CCC1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 131°C to 133°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.502
Quantity 50 g
Beilstein 2725083
Formula Weight 206.24
Percent Purity ≥90%
Chemical Name or Material Ethyl 2-oxo-4-phenylbutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.