missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxo-4-phenylbutyrate (>90%), TRC

Product Code. 30375962
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375962 10 g 10g
30371744 100 g -
30373246 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375962 Supplier Toronto Research Chemicals Supplier No. TRCE92538510G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64920-29-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
Synonym alpha-Oxo-benzenebutanoic Acid Ethyl Ester-d5, 2-Oxo-4-phenylbutyric Acid Ethyl Ester, 4-Phenyl-2-oxobutyric Acid Ethyl Ester, Ethyl 2-Oxo-4-phenylbutanoate, Ethyl 4-Phenyl-2-oxobutanoate, Ethyl 4-Phenyl-2-oxobutyrate, Ethyl Benzylpyruvate
IUPAC Name ethyl 2-oxo-4-phenylbutanoate
SMILES CCOC(=O)C(=O)CCc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H14O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3
Formula Weight 206.0943
Chemical Name or Material Ethyl 2-oxo-4-phenylbutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.