missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(3,4,5-trimethoxyphenyl)propionate, 98%, Thermo Scientific™

Product Code. 11459693
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459693 25 g 25g
11449693 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459693 Supplier Thermo Scientific Alfa Aesar Supplier No. A12459.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70311-20-5
Molecular Formula C14H20O5
Molecular Weight (g/mol) 268.309
MDL Number MFCD00017522
InChI Key AYBWQZGGKFGQLB-UHFFFAOYSA-N
Synonym ethyl 3-3,4,5-trimethoxyphenyl propionate, ethyl 3-3,4,5-trimethoxyphenyl propanoate, ethyl 3,4,5-trimethoxy dihydrocinnamate, ethyl 3-3,4,5-trimethoxyphenyl propanoate #, 3,4,5-trimethoxybenzenepropionic acid ethyl ester, 3-3,4,5-trimethoxyphenyl propionic acid ethyl ester, benzenepropanoic acid, 3,4,5-trimethoxy-, ethyl ester
PubChem CID 622665
IUPAC Name ethyl 3-(3,4,5-trimethoxyphenyl)propanoate
SMILES CCOC(=O)CCC1=CC(=C(C(=C1)OC)OC)OC

Specifications

Quantity 25 g
Formula Weight 268.31
Percent Purity 98%
Chemical Name or Material Ethyl 3- (3,4,5-trimethoxyphenyl)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.