missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-dimethylaminopropionate, 97%

Product Code. 11361317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361317 25 g 25g
11351317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361317 Supplier Thermo Scientific Alfa Aesar Supplier No. L01542.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20120-21-2 | C7H15NO2 | 145.20 g/mol

Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-dimethylaminopropionate is used in the preparation of [3-(dimethylamino)propionyl]guanidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20120-21-2
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.20
MDL Number MFCD00015166
InChI Key IZMJWGJGVGRWBW-UHFFFAOYSA-N
Synonym ethyl 3-dimethylamino propanoate, ethyl 3-dimethylaminopropionate, ethyl 3-dimethylamino propionate, n,n-dimethyl-beta-alanine ethyl ester, ethyl 3-n,n-dimethylamino propionate, ethyl n,n-dimethyl-beta-alaninate, ethyl beta-dimethylamino propionate, beta-alanine, n,n-dimethyl-, ethyl ester, ethyl 3-dimethyl-aminopropionate, ethyl-3-n,n-dimethylamino propionate
PubChem CID 88372
IUPAC Name ethyl 3-(dimethylamino)propanoate
SMILES CCOC(=O)CCN(C)C

Specifications

Density 0.915
Boiling Point 56°C to 57°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Amine-like
Refractive Index 1.419
Quantity 25 g
UN Number UN1993
Beilstein 1702477
Solubility Information Immiscible with water.
Formula Weight 145.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-dimethylaminopropionate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 3-dimethylaminopropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl 3-dimethylaminopropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.