missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Ethoxy-2-Propenoate, 98%, BLD Pharm™

Product Code. 31031190
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
1g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31031190 500 g -
31028019 1 g 1g
31025690 100 g -
31014806 5 g -
31025744 25 g -
31024015 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31031190 Supplier BLD Pharm Supplier No. BD128636500g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1001-26-9
Molecular Formula C7H12O3
MDL Number MFCD00009863
Quantity 500 g
SMILES O=C(OCC)/C=C/OCC
Molecular Weight (g/mol) 144.17
Percent Purity 98%
Chemical Name or Material Ethyl 3-Ethoxy-2-Propenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.