missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-3-nitrobenzoate, 97%, Thermo Scientific™

Product Code. 10352185
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10352185 100 g Amber Glass Bottle 100g
10618765 25 g Amber Glass Bottle 25g
10149829 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10352185 Supplier Thermo Scientific Maybridge Supplier No. JP00516FA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16588-16-2
Molecular Formula C9H8ClNO4
Molecular Weight (g/mol) 229.616
InChI Key BLNLZRQIUGDTAO-UHFFFAOYSA-N
Synonym benzoic acid, 4-chloro-3-nitro-, ethyl ester, 4-chloro-3-nitrobenzoic acid ethyl ester, 4-chloro-3-nitro-benzoic acid ethyl ester, ethyl 4-chloranyl-3-nitro-benzoate, acmc-20a7tu, ethyl4-chloro-3-nitrobenzoate, ethyl 4-chloro-3-nitro-benzoate, ethyl-4-chloro-3-nitro-benzoate, ethyl-4-chloro-3-nitrobenzoate, 3-nitro-4-chlorobenzoic acid ethyl ester
PubChem CID 1268247
IUPAC Name ethyl 4-chloro-3-nitrobenzoate
SMILES CCOC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 100 g
Formula Weight 229.62
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-3-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.