Learn More
Ethyl 4-chloro-7-methoxyquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
Product Code. 15425046
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425046 1 g 1g
15435046 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15425046 Supplier Thermo Scientific Alfa Aesar Supplier No. H59473.03
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77156-85-5
Molecular Formula C13H12ClNO3
Molecular Weight (g/mol) 265.693
MDL Number MFCD02217815
InChI Key IXMRUGGFGJIANG-UHFFFAOYSA-N
Synonym ethyl 4-chloro-7-methoxy-3-quinolinecarboxylate, 4-chloro-7-methoxyquinoline-3-carboxylic acid ethyl ester, cbmicro_031264, ethyl 4-chloro-7-methoxyquinoline-3-carboxylate, 4-chloro-7-methoxy-quinoline-3-carboxylic acid ethyl ester, 4-chloro-3-ethoxycarbonyl-7-methoxyquinoline, 7-methoxy-4-chloro-3-ethoxycarbonylquinoline, ethyl 4-chloro-7-methoxy-quinoline-3-carboxylate
PubChem CID 728976
IUPAC Name ethyl 4-chloro-7-methoxyquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=C(C=CC2=C1Cl)OC
Specifications
Quantity 1 g
Formula Weight 265.7
Percent Purity 97%
Chemical Name or Material Ethyl 4-chloro-7-methoxyquinoline-3-carboxylate