missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Chloroacetoacetate, TRC

Product Code. 30368814
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368814 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368814 Supplier Toronto Research Chemicals Supplier No. TRCE90180010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-Chloroacetoacetate
CAS 638-07-3
Molecular Formula C6 H9 Cl O3
Quantity 10 g
Synonym Butanoic acid, 4-chloro-3-oxo-, ethyl ester, Acetoacetic acid, 4-chloro-, ethyl ester (7CI,8CI), 4-Chloro acetoethylacetate, 4-Chloro-3-oxobutanoic acid ethyl ester, 4-Chloro-3-oxobutyric acid ethyl ester, 4-Chloroacetoacetic acid ethyl ester, Ethyl (chloroacetyl)acetate, Ethyl 3-oxo-4-chlorobutanoate, Ethyl 4-chloro-3-oxobutanoate, Ethyl 4-chloro-3-oxobutyrate, Ethyl 4-chloroacetoacetate, Ethyl chloroacetoacetate, Ethyl γ-chloroacetoacetate, Ethyl ω-chloroacetoacetate, γ-Chloroacetoacetic acid ethyl ester
InChI Formula InChI=1S/C6H9ClO3/c1-2-10-6(9)3-5(8)4-7/h2-4H2,1H3
SMILES CCOC(=O)CC(=O)CCl
IUPAC Name ethyl 4-chloro-3-oxobutanoate
Molecular Weight (g/mol) 164.59
Formula Weight 164.024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.