missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Dimethylaminobenzoate, TRC

Product Code. 30382398
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382398 Supplier Toronto Research Chemicals Supplier No. TRCE9166201G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Dimethylaminobenzoic acid ethyl ester
CAS 10287-53-3
Purity Grade Notes HPLC
Molecular Formula C11 H15 N O2
Quantity 1 g
Synonym Benzoic acid, p-(dimethylamino)-, ethyl ester (6CI,7CI,8CI), 4-Dimethylaminobenzoic acid ethyl ester, 4-Ethoxycarbonyl-N,N-dimethylaniline, Darocur EBD, Darocur EDB, EDAB, EDB, EDMAB, EPD, Ethyl 4-(N,N-dimethylamino)benzoate, Ethyl 4-(dimethylamino)benzoate, Ethyl N,N-dimethylamino-p-benzoate, Ethyl p-(N,N-dimethylamino)benzoate, Ethyl p-(dimethylamino)benzoate, Genocure EPD, IHT-PI EDB, JRCure 1101, Kayacure EPA, Luna EDB, Lunacure EDB, N,N-Dimethylbenzocaine, Omnirad EDB, PI-EDB, Parbenate, Photoinitiator EDAB, Quantacure EPD, SB-PI 704, Speedcure EDB, p-(Ethoxycarbonyl)-N,N-dimethylaniline, π-EDB
InChI Formula InChI=1S/C11H15NO2/c1-4-14-11(13)9-5-7-10(8-6-9)12(2)3/h5-8H,4H2,1-3H3
SMILES CCOC(=O)c1ccc(cc1)N(C)C
IUPAC Name ethyl 4-(dimethylamino)benzoate
Molecular Weight (g/mol) 193.24
Formula Weight 193.1103
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.