missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide (Piroxicam Impurity K), TRC

Product Code. 17953695
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953695 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17953695 Supplier Toronto Research Chemicals Supplier No. TRCE9189805G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24683-26-9
Molecular Formula C12 H13 N O5 S
Quantity 5 g
Synonym Ethyl 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Meloxicam Imp. A (EP), Piroxicam Imp. K (EP)
InChI Formula InChI=1S/C12H13NO5S/c1-3-18-12(15)10-11(14)8-6-4-5-7-9(8)19(16,17)13(10)2/h4-7,14H,3H2,1-2H3
SMILES CCOC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1C
IUPAC Name ethyl 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxylate
Molecular Weight (g/mol) 283.3
Formula Weight 283.0514
Percent Purity >95%
Chemical Name or Material ethyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-dioxide
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.