missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-iodobenzoate, 98%

Product Code. 11481958
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481958 250 g 250g
11461958 10 g 10g
11471958 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481958 Supplier Thermo Scientific Alfa Aesar Supplier No. A14570.30
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51934-41-9 | C9H9IO2 | 276.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51934-41-9
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD00017344
InChI Key YCBJOQUNPLTBGG-UHFFFAOYSA-N
Synonym p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
PubChem CID 142891
IUPAC Name ethyl 4-iodobenzoate
SMILES CCOC(=O)C1=CC=C(I)C=C1

Specifications

Density 1.66
Boiling Point 283°C
Flash Point >110°C (230°F)
Refractive Index 1.588
Quantity 250 g
Beilstein 2208568
Sensitivity Light sensitive
Formula Weight 276.08
Percent Purity 98%
Chemical Name or Material Ethyl 4-iodobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.