missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate, 98%

Product Code. 11435413
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435413 25 g 25g
11415413 1 g 1g
11425413 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435413 Supplier Thermo Scientific Alfa Aesar Supplier No. A13399.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18212-20-9 | C6H8N2O2S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18212-20-9
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00052549
InChI Key AHPXTXGCMLOXGA-UHFFFAOYSA-N
Synonym ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid ethylester, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-, ethyl ester, acmc-209xfp, ethylmethylthiadiazolecarboxylate, 5-ethoxycarbonyl-4-methyl-1,2,3-thiadiazole, 4-methyl-5-thiadiazolecarboxylic acid ethyl ester, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid ethyl ester, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid ethyl ester
PubChem CID 2758878
IUPAC Name ethyl 4-methylthiadiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=NS1

Specifications

Density 1.265
Boiling Point 117°C to 118°C (15 mmHg)
Refractive Index 1.504
Quantity 25 g
Beilstein 609140
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.