missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 15491666
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491666 250 mg 250mg
15471666 1 g 1g
15481666 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491666 Supplier Thermo Scientific Alfa Aesar Supplier No. H54842.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383146-88-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.30
MDL Number MFCD01936157
InChI Key UGCVLLXXBVTJPK-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-pyridin-2-yl thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-2-ylthiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-2-pyridyl thiazole-5-carboxylate, ethyl4-methyl-2-pyridin-2-yl thiazole-5-carboxylate
PubChem CID 2763623
IUPAC Name ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=C(S1)C1=CC=CC=N1

Specifications

Quantity 250 mg
Formula Weight 248.3
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.