missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]thiazole-5-carboxylate, 97%, Alfa Aesar™

Product Code. 15490886
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490886 250 mg 250mg
15470886 1 g 1g
15480886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490886 Supplier Thermo Scientific Alfa Aesar Supplier No. H54202.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-03-9
Molecular Formula C14H12F3NO2S
Molecular Weight (g/mol) 315.31
MDL Number MFCD00068104
InChI Key LPIXRQSYBTUXOQ-UHFFFAOYSA-N
PubChem CID 2737210
IUPAC Name ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=CC=C(C=C2)C(F)(F)F)C

Specifications

Quantity 250 mg
Beilstein 9409771
Synonym ethyl 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxylate, 4-methyl-2-4-trifluoromethylphenyl thiazole-5-carboxylic acid ethyl ester, ethyl 4-methyl-2-4-trifluoromethyl phenyl-thiazole-5-carboxylate, 4-methyl-2-4-trifluoromethyl-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 4-methyl-2-4-trifluoromethyl phenyl-, ethyl ester, ethyl4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxylate, ethyl 4-methyl-2-4-trifluoromethyl phenyl thiazo
Formula Weight 315.31
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.