missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitrocinnamate, 99%

Product Code. 11464614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464614 100 g 100g
11444614 5 g 5g
11454614 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464614 Supplier Thermo Scientific Alfa Aesar Supplier No. A13196.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 953-26-4 | C11H11NO4 | 221.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953-26-4
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.212
MDL Number MFCD00007380
InChI Key PFBQVGXIMLXCQB-VMPITWQZSA-N
Synonym ethyl 4-nitrocinnamate, ethyl p-nitrocinnamate, ethyl 3-4-nitrophenyl acrylate, 2-propenoic acid, 3-4-nitrophenyl-, ethyl ester, cinnamic acid, p-nitro-, ethyl ester, ethyl-4-nitrocinnamate, e-ethyl 3-4-nitrophenyl acrylate, propenoic acid, 3-4-nitrophenyl-, ethyl ester, 4-nitrozimtsaeure-ethylester, ethyl 2e-3-4-nitrophenyl prop-2-enoate
PubChem CID 1268038
IUPAC Name ethyl (E)-3-(4-nitrophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 138°C to 140°C
Quantity 100 g
Beilstein 2052533
Formula Weight 221.21
Percent Purity 99%
Chemical Name or Material Ethyl 4-nitrocinnamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.