missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 11339989
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339989 1 g 1g
11329989 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339989 Supplier Thermo Scientific Alfa Aesar Supplier No. H25764.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate is used in the preparation of fused 1,3,6- thiadiazepines such as di[1,2,3]triazolo[1,5-a:5',1'-d][3,1,5]benzothiadiazepines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate is used in the preparation of fused 1,3,6- thiadiazepines such as di[1,2,3]triazolo[1,5-a:5′,1′-d][3,1,5]benzothiadiazepines.

Solubility
Miscible with dimethyl formamide.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6439-91-4
Molecular Formula C5H5BrN2O2S
Molecular Weight (g/mol) 237.071
MDL Number MFCD02324685
InChI Key MBEUYMRLYKXBGT-UHFFFAOYSA-N
Synonym ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 5-bromo-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, acmc-209wgk, 1,2,3-thiadiazole-4-carboxylicacid, 5-bromo-, ethyl ester, 5-bromo-1,2,3-thiadiazole-4-carboxylic acid ethyl
PubChem CID 11020903
IUPAC Name ethyl 5-bromothiadiazole-4-carboxylate
SMILES CCOC(=O)C1=C(SN=N1)Br

Specifications

Melting Point 76°C to 78°C
Quantity 1 g
Solubility Information Miscible with dimethyl formamide.
Formula Weight 237.07
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.