missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (5-Ethyl-2-pyridinyl)acetate, TRC

Product Code. 30382507
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382507 5 g
30372718 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382507 Supplier Toronto Research Chemicals Supplier No. TRCE9179055G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99986-02-4
Molecular Formula C11 H15 N O2
Molecular Weight (g/mol) 193.24
Synonym 2-Pyridineacetic acid, 5-ethyl-, ethyl ester, 5-Ethyl-2-pyridineacetic acid ethyl ester, Ethyl (5-ethyl-2-pyridinyl)acetate
IUPAC Name ethyl 2-(5-ethylpyridin-2-yl)acetate
SMILES CCOC(=O)Cc1ccc(CC)cn1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H15NO2/c1-3-9-5-6-10(12-8-9)7-11(13)14-4-2/h5-6,8H,3-4,7H2,1-2H3
Formula Weight 193.1103
Chemical Name or Material Ethyl (5-Ethyl-2-pyridinyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.