missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(thiophen-2-yl)isoxazole-3-carboxylate, TRC

Product Code. 30386579
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386579 250 mg
30373461 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386579 Supplier Toronto Research Chemicals Supplier No. TRCE927670250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90924-54-2
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
IUPAC Name ethyl 5-thiophen-2-yl-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)c1cc(on1)c2cccs2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H9NO3S/c1-2-13-10(12)7-6-8(14-11-7)9-4-3-5-15-9/h3-6H,2H2,1H3
Formula Weight 223.0303
Chemical Name or Material Ethyl 5-(thiophen-2-yl)isoxazole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.