Learn More
enQuireBio™ Recombinant Human ATP6V1F Protein
Product Code. p-7155609
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16017621 10 μg 10µg
16057621 50 μg 50µg
16007621 100 μg 100µg
16037621 200 μg 200µg
16047621 500 μg 500µg
16027621 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16017621 Supplier enQuireBio™ Supplier No. QP90910ug
Add to basket
Add to basket
A DNA sequence encoding the Homo sapiens (Human) ATP6V1F, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001185838.1
Gene ID (Entrez) 9296
Name ATP6V1F Protein
Quantity 10 μg
Regulatory Status RUO
Endotoxin Concentration Not determined.
Gene Symbol ATP6V1F
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MAGRGKLIAV IGDEDTVTGF LLGGIGELNK NRHPNFLVVE KDTTINEIED TFRQFLNRDD IGIILINQYI AEMVRHALDA HQQSIPAVLE IPSKEHPYDA AKDSILRRAR GMFTAEDLR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.