Learn More
Ethyl 7-Chloro-2-oxoheptanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 7-Chloro-2-oxoheptanoate |
| CAS | 78834-75-0 |
| Molecular Formula | C9H15ClO3 |
| Quantity | 5 g |
| Synonym | 7-Chloro-2-oxo-heptanoic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C9H15ClO3/c1-2-13-9(12)8(11)6-4-3-5-7-10/h2-7H2,1H3 |
| SMILES | CCOC(=O)C(=O)CCCCCCl |
| Molecular Weight (g/mol) | 206.67 |
| Formula Weight | 206.071 |