missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, 97%, Thermo Scientific™

Product Code. p-2704964
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10009091 1 g Amber Glass Bottle 1g
10282722 10 g Amber Glass Bottle 10g
11370517 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10009091 Supplier Thermo Scientific Maybridge Supplier No. BTB02003DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 71083-06-2
Molecular Formula C12H10FNO3
Molecular Weight (g/mol) 235.214
MDL Number MFCD00052242
InChI Key MPUYCZQHTGRPNE-UHFFFAOYSA-N
Synonym ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate, ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate, 3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester, 8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester, 8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester, pubchem19599, maybridge1_000540, acmc-209nc3
PubChem CID 707160
IUPAC Name ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
SMILES CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.21
Percent Purity 97%
Chemical Name or Material Ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.