missing translation for 'onlineSavingsMsg'
Learn More

Ethyl b-D-Glucuronide (1 mg/mL in Methanol), TRC

Product Code. 30473977
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473977 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473977 Supplier Toronto Research Chemicals Supplier No. TRCKIT83201X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl-beta-D-glucuronide
CAS 17685-04-0
Molecular Formula C8 H14 O7
Quantity 1x1 mL
Synonym β-D-Glucopyranosiduronic acid, ethyl (9CI, ACI), Ethyl β-D-glucopyranosiduronic acid (ACI), Glucopyranosiduronic acid, ethyl, β-D- (6CI, 8CI), BA 2829, Ethyl glucuronide, Ethyl β-D-glucosiduronic acid, Ethyl β-D-glucuronide
InChI Formula InChI=1S/C8H14O7/c1-2-14-8-5(11)3(9)4(10)6(15-8)7(12)13/h3-6,8-11H,2H2,1H3,(H,12,13)/t3-,4-,5+,6-,8+/m0/s1
SMILES CCO[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6R)-6-ethoxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 222.193
Formula Weight 222.074
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.