missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-Beta-D-glucuronide, Lipomed

Product Code. 30458563
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458563 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458563 Supplier Lipomed Supplier No. LPMEGL33210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Ethyl-Beta-D-glucuronide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl-beta-D-glucuronide
CAS 17685-04-0
Analyte or Component Names Ethyl-beta-D-glucuronide
Shipping Condition Room Temperature
Molecular Formula C8 H14 O7
Synonym β-D-Glucopyranosiduronic acid, ethyl (9 CI, ACI), Ethyl β-D-glucopyranosiduronic acid (ACI), Glucopyranosiduronic acid, ethyl, β-D- (6 CI, 8 CI), BA 2829, Ethyl glucuronide, Ethyl β-D-glucosiduronic acid, Ethyl β-D-glucuronide
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CCO[C@@H]1 O[C@@H]([C@@H](O)[C@H](O)[C@H]1 O)C(=O)O
IUPAC Name (2 S,3 S,4 S,5 R,6 R)-6-ethoxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 222.193
Quantity 10 mg
Formula Weight 222.074 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.