missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Beta-D-Glucuronide, TRC

Product Code. 17960555
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960555 10 mg 10mg
17931185 1 mg 1mg
17922245 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960555 Supplier Toronto Research Chemicals Supplier No. TRCE91842510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17685-04-0
Molecular Formula C8 H14 O7
Molecular Weight (g/mol) 222.193
Synonym β-D-Glucopyranosiduronic acid, ethyl (9CI, ACI), Ethyl β-D-glucopyranosiduronic acid (ACI), Glucopyranosiduronic acid, ethyl, β-D- (6CI, 8CI), BA 2829, Ethyl glucuronide, Ethyl β-D-glucosiduronic acid, Ethyl β-D-glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-6-ethoxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CCO[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H14O7/c1-2-14-8-5(11)3(9)4(10)6(15-8)7(12)13/h3-6,8-11H,2H2,1H3,(H,12,13)/t3-,4-,5+,6-,8+/m0/s1
Formula Weight 222.074
Chemical Name or Material Ethyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.