missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromodifluoroacetate, 97%

Product Code. 11434437
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434437 1 g 1g
11444437 5 g 5g
11454437 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434437 Supplier Thermo Scientific Alfa Aesar Supplier No. B20461.03
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 667-27-6 | C4H5BrF2O2 | 202.983 g/mol

Ethyl bromodifluoroacetate, is used for the treatment of the Reformat sky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. It is also used in the analysis of reagent purity through IR, NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromodifluoroacetate, is used for the treatment of the Reformat sky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. It is also used in the analysis of reagent purity through IR, NMR.

Solubility
Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 667-27-6
Molecular Formula C4H5BrF2O2
Molecular Weight (g/mol) 202.983
MDL Number MFCD00042069
InChI Key IRSJDVYTJUCXRV-UHFFFAOYSA-N
Synonym ethyl bromodifluoroacetate, ethylbromodifluoroacetate, bromodifluoroacetic acid ethyl ester, acetic acid, bromodifluoro-, ethyl ester, ethyl difluorobromoacetate, ethyl 2-bromo-2,2-difluoro-acetate, 2-bromo-2,2-difluoro-1-ethoxyethan-1-one, 2-bromo-2,2-difluoro-1-ethoxy-1-oxoethane, ethylbrom difluor acetat, pubchem16953
PubChem CID 69585
IUPAC Name ethyl 2-bromo-2,2-difluoroacetate
SMILES CCOC(=O)C(F)(F)Br

Specifications

Boiling Point 112°C to 114°C
Flash Point 21°C (69°F)
Quantity 1 g
UN Number UN2924
Beilstein 1906095
Formula Weight 202.99
Percent Purity 97%
Chemical Name or Material Ethyl bromodifluoroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.