missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Bromodifluoroacetate, TRC

Product Code. 30377210
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377210 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377210 Supplier Toronto Research Chemicals Supplier No. TRCE67845010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Bromodifluoroacetate
CAS 667-27-6
Molecular Formula C4H5BrF2O2
Quantity 10 g
Synonym 2-Bromo-2,2-difluoroacetic Acid Ethyl Ester, Bromodifluoroacetic Acid Ethyl Ester, Ethyl 2-Bromo-2,2-difluoroacetate, Ethyl Difluorobromoacetate
InChI Formula InChI=1S/C4H5BrF2O2/c1-2-9-3(8)4(5,6)7/h2H2,1H3
SMILES CCOC(=O)C(F)(F)Br
IUPAC Name ethyl 2-bromo-2,2-difluoroacetate
Molecular Weight (g/mol) 202.98
Formula Weight 201.9441
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.