missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chlorodifluoroacetate, 98%

Product Code. 11422347
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422347 25 g 25g
11432347 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422347 Supplier Thermo Scientific Alfa Aesar Supplier No. A14232.14
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 383-62-0 | C4H5ClF2O2 | 158.529 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383-62-0
Molecular Formula C4H5ClF2O2
Molecular Weight (g/mol) 158.529
MDL Number MFCD00013662
InChI Key GVCAWQUJCHZRCB-UHFFFAOYSA-N
Synonym ethyl chlorodifluoroacetate, chlorodifluoroacetic acid ethyl ester, ethyl chlorodifluoroethanoate, acetic acid, chlorodifluoro-, ethyl ester, ethylchlorodifluoroacetate, acetic acid, 2-chloro-2,2-difluoro-, ethyl ester, ethyl 2-chloro-2,2-difluoro-acetate, pubchem12573, ethyl chlordifluoroacetate, acmc-1cr8n
PubChem CID 67843
IUPAC Name ethyl 2-chloro-2,2-difluoroacetate
SMILES CCOC(=O)C(F)(F)Cl

Specifications

Density 1.252
Boiling Point 96°C to 97°C
Flash Point 18°C (64°F)
Refractive Index 1.358
Quantity 25 g
UN Number UN2924
Beilstein 1761413
Sensitivity Moisture sensitive
Formula Weight 158.53
Percent Purity 98%
Chemical Name or Material Ethyl chlorodifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.