missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Difluoroacetate, TRC

Product Code. 30371031
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30371031 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30371031

Brand: Toronto Research Chemicals TRCE91660310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Difluoroacetate
CAS 454-31-9
Molecular Formula C4H6F2O2
Quantity 10 g
InChI Formula InChI=1S/C4H6F2O2/c1-2-8-4(7)3(5)6/h3H,2H2,1H3
SMILES CCOC(=O)C(F)F
Molecular Weight (g/mol) 124.09
Formula Weight 124.0336
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.