missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Glycolate, TRC

Product Code. 30375191
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30375191 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30375191

Brand: Toronto Research Chemicals TRCE91854010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Glycolate
CAS 623-50-7
Molecular Formula C4H8O3
Quantity 10 g
Synonym Ethyl 2-Hydroxyacetate, Ethyl Hydroxyacetate, Glycolic Acid Ethyl Ester, 2-Hydroxyacetic Acid Ethyl Ester, NSC 8835
InChI Formula InChI=1S/C4H8O3/c1-2-7-4(6)3-5/h5H,2-3H2,1H3
SMILES CCOC(=O)CO
IUPAC Name ethyl 2-hydroxyacetate
Molecular Weight (g/mol) 104.1
Formula Weight 104.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.