missing translation for 'onlineSavingsMsg'
Learn More

Ethyl homovanillate, 97%, Thermo Scientific™

Product Code. 10284594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10284594 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10284594 Supplier Thermo Scientific Acros Supplier No. 189240010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 60563-13-5
Melting Point 44°C to 47°C
Boiling Point 180°C to 185°C (14.0 mmHg)
Flash Point 113°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C11H14O4
Linear Formula HOC6H3(OCH3)CH2CO2C2H5
MDL Number MFCD00009180
Quantity 1 g
Beilstein 10,I,198
Synonym ethyl homovanillate, ethyl 2-4-hydroxy-3-methoxyphenyl acetate, ethyl 4-hydroxy-3-methoxyphenylacetate, 4-hydroxy-3-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-hydroxy-3-methoxy-, ethyl ester, benzeneacetic acid,4-hydroxy-3-methoxy-, ethyl ester, acmc-20akxa, awpmwzhwvkxadv-uhfffaoysa, ethyl 3-methoxy-4-hydroxy-phenylacetate
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol and acetone
InChI Key AWPMWZHWVKXADV-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=CC=C(O)C(OC)=C1
IUPAC Name ethyl 2-(4-hydroxy-3-methoxyphenyl)acetate
Molecular Weight (g/mol) 210.23
PubChem CID 108965
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material Ethyl homovanillate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl homovanillate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.