missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Indole-3-carboxylate, Mikromol™

Product Code. 30462033
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30462033

Brand: Mikromol MM0763.0300254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Indole-3-carboxylate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl indole-3-carboxylate
CAS 776-41-0
Analyte or Component Names Ethyl indole-3-carboxylate
Shipping Condition Room Temperature
Molecular Formula C11 H11 N O2
Certifications/Compliance ISO 17025
Synonym Ethyl Indole-3-carboxylate, 1H-Indole-3-carboxylic acid ethyl ester, Indole-3-carboxylic acid ethyl ester, 3-(Ethoxycarbonyl)indole, Ethyl 1H-indole-3-carboxylate, Ethyl indole-3-carboxylate, NSC 63796
InChI Formula InChI=1S/C11H11NO2/c1-2-14-11(13)9-7-12-10-6-4-3-5-8(9)10/h3-7,12H,2H2,1H3
Solution Type Neat
SMILES CCOC(=O)c1c[nH]c2ccccc12
IUPAC Name ethyl 1H-indole-3-carboxylate
Molecular Weight (g/mol) 189.21
Quantity 4 x 25 mg
Formula Weight 189.079 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.